Assay Details
| Gene Symbol: EGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097488.1 | NP_001090957.1 | 1 - 2 | 250 | 71 | |
| XM_013983286.2 | XP_013838740.1 | 338 | 71 | |||
| GenBank mRNA | AK344043.1 | - | 1 - 2 | 189 | 71 | |
| DQ665829.1 | - | 1 - 2 | 250 | 71 | ||
More Information
Set Membership: |
|

EGR2
Genomic Map