Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097492.1 | NP_001090961.1 | 4 - 5 | 908 | 90 | |
| GenBank mRNA | AB189912.1 | - | 4 - 5 | 908 | 90 | |
| AK352055.1 | - | 4 - 5 | 1034 | 90 | ||
More Information
Set Membership: |
|

CD1E
Genomic Map