Assay Details
| Gene Symbol: NR6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097499.1 | NP_001090968.1 | 7 - 8 | 1243 | 66 | |
| XM_021098706.1 | XP_020954365.1 | 1293 | 66 | |||
| XM_021099196.1 | XP_020954855.1 | 2859 | 66 | |||
| XM_021099647.1 | XP_020955306.1 | 2856 | 66 | |||
| GenBank mRNA | AB248749.1 | - | 7 - 8 | 1243 | 66 | |
More Information
Set Membership: |
|

NR6A1
Genomic Map