Assay Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097501.2 | NP_001090970.1 | 2 - 3 | 377 | 63 | |
| XM_005659974.3 | XP_005660031.1 | 509 | 63 | |||
| GenBank mRNA | AK232068.1 | - | 3 - 4 | 469 | 63 | |
| AK350128.1 | - | 3 - 4 | 437 | 63 | ||
| AK352387.1 | - | 3 - 4 | 442 | 63 | ||
| EF053388.1 | - | 2 - 3 | 380 | 63 | ||
More Information
Set Membership: |
|

LGALS3
Genomic Map