Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097516.1 | NP_001090985.1 | 2 - 3 | 169 | 81 | |
| GenBank mRNA | AK238737.1 | - | 2 - 3 | 193 | 81 | |
| AK346167.1 | - | 2 - 3 | 172 | 81 | ||
| AK400044.1 | - | 2 - 3 | 229 | 81 | ||
| AY682219.1 | - | 2 - 3 | 200 | 81 | ||
| AY682222.1 | - | 2 - 3 | 200 | 81 | ||
| AY682832.1 | - | 2 - 3 | 186 | 81 | ||
| DQ486897.1 | - | 2 - 3 | 169 | 81 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map