Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099923.1 | NP_001093393.1 | 1 - 2 | 333 | 68 | |
| GenBank mRNA | AB292176.1 | - | 1 - 2 | 370 | 68 | |
| EF198416.1 | - | 1 - 2 | 333 | 68 | ||
| EU056736.1 | - | 1 - 2 | 382 | 68 | ||
| EU077229.1 | - | 1 - 2 | 326 | 68 | ||
| EU155133.1 | - | 1 - 2 | 156 | 68 | ||
More Information
Set Membership: |
|

MYD88
Genomic Map