Assay Details
| Gene Symbol: SOX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099933.1 | NP_001093403.1 | 3 - 4 | 821 | 91 | |
| XM_013997513.2 | XP_013852967.2 | 1114 | 91 | |||
| GenBank mRNA | AB271933.1 | - | 3 - 4 | 821 | 91 | |
More Information
Set Membership: |
|

SOX10
Genomic Map