Assay Details
| Gene Symbol: SQLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101026.1 | NP_001094496.1 | 4 - 5 | 1294 | 67 | |
| XM_005662853.3 | XP_005662910.1 | 1732 | 67 | |||
| GenBank mRNA | AB530156.1 | - | 4 - 5 | 828 | 67 | |
| AK233408.1 | - | 4 - 5 | 1710 | 67 | ||
| AK394121.1 | - | 4 - 5 | 1710 | 67 | ||
| EF683117.1 | - | 4 - 5 | 1294 | 67 | ||
More Information
Set Membership: |
|

SQLE
Genomic Map