Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101815.1 | NP_001095285.1 | 6 - 7 | 887 | 102 | |
| GenBank mRNA | AK233128.1 | - | 6 - 7 | 857 | 102 | |
| AK346370.1 | - | 6 - 7 | 947 | 102 | ||
| AK394195.1 | - | 6 - 7 | 967 | 102 | ||
| AY349422.1 | - | 6 - 7 | 876 | 102 | ||
More Information
Set Membership: |
|

C2
Genomic Map