Assay Details
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101820.1 | NP_001095290.1 | 8 - 9 | 672 | 81 | |
XM_013985457.2 | XP_013840911.1 | 631 | 81 | |||
GenBank mRNA | AB529457.1 | - | 8 - 9 | 672 | 81 | |
AK231734.1 | - | 8 - 9 | 685 | 81 | ||
AK398900.1 | - | 8 - 9 | 683 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
