Assay Details
| Gene Symbol: NEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101822.1 | NP_001095292.1 | 3 - 4 | 752 | 80 | |
| GenBank mRNA | EU282367.1 | - | 3 - 4 | 618 | 80 | |
More Information
Set Membership: |
|

NEU1
Genomic Map