Assay Details
| Gene Symbol: BCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103213.2 | NP_001096683.1 | 1 - 2 | 75 | 132 | |
| GenBank mRNA | AK393206.1 | - | 1 - 2 | 282 | 133 | |
| EU088132.1 | - | 1 - 2 | 76 | 132 | ||
| FJ376731.1 | - | 1 - 2 | 139 | 132 | ||
More Information
Set Membership: |
|

BCL10
Genomic Map