Assay Details
| Gene Symbol: TOR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105298.1 | NP_001098768.1 | 4 - 5 | 749 | 95 | |
| GenBank mRNA | AK233486.1 | - | 4 - 5 | 810 | 95 | |
| EF101283.1 | - | 4 - 5 | 749 | 95 | ||
| EF101284.1 | - | 4 - 5 | 749 | 95 | ||
More Information
Set Membership: |
|

TOR1A
Genomic Map