Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109947.1 | NP_001103417.1 | 1 - 2 | 829 | 68 | |
| XM_005667787.3 | XP_005667844.1 | 5076 | 68 | |||
| GenBank mRNA | EF154830.1 | - | 1 - 2 | 829 | 68 | |
More Information
Set Membership: |
|

ANGPTL1
Genomic Map