Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110174.1 | NP_001103644.1 | 5 - 6 | 1181 | 55 | |
| XM_013985484.2 | XP_013840938.1 | 1196 | 55 | |||
| XM_013985485.2 | XP_013840939.1 | 1196 | 55 | |||
More Information
Set Membership: |
|

CTCFL
Genomic Map