Assay Details
| Gene Symbol: GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110419.2 | NP_001103889.2 | 2 - 3 | 375 | 66 | |
| GenBank mRNA | AK234318.1 | - | 2 - 3 | 370 | 66 | |
| AK346447.1 | - | 2 - 3 | 370 | 66 | ||
| AK397541.1 | - | 2 - 3 | 376 | 66 | ||
| BP144649.1 | - | 2 - 3 | 386 | 66 | ||
| D13308.1 | - | 2 - 3 | 323 | 66 | ||
More Information
Set Membership: |
|

GNMT
Genomic Map