Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110428.1 | NP_001103898.1 | 3 - 4 | 523 | 57 | |
| XM_021097865.1 | XP_020953524.1 | 539 | 57 | |||
| XM_021097866.1 | XP_020953525.1 | 647 | 57 | |||
| GenBank mRNA | AK234305.1 | - | 3 - 4 | 497 | 57 | |
| AK238459.1 | - | 3 - 4 | 453 | 57 | ||
| AY610363.1 | - | 3 - 4 | 403 | 57 | ||
More Information
Set Membership: |
|

CNPY3
Genomic Map