Assay Details
| Gene Symbol: RTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110432.1 | NP_001103902.1 | 6 - 7 | 688 | 76 | |
| XM_005673032.3 | XP_005673089.1 | 675 | 76 | |||
| GenBank mRNA | AK235189.1 | - | 6 - 7 | 661 | 76 | |
| AK348007.1 | - | 6 - 7 | 614 | 76 | ||
| AK397035.1 | - | 6 - 7 | 664 | 76 | ||
| AK397163.1 | - | 6 - 7 | 663 | 76 | ||
| AK399170.1 | - | 6 - 7 | 663 | 76 | ||
More Information
Set Membership: |
|

RTF2
Genomic Map