Assay Details
| Gene Symbol: RTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110432.1 | NP_001103902.1 | 8 - 9 | 846 | 57 | |
| GenBank mRNA | AK235189.1 | - | 8 - 9 | 819 | 57 | |
| AK348007.1 | - | 8 - 9 | 772 | 57 | ||
| AK397163.1 | - | 8 - 9 | 821 | 57 | ||
More Information
Set Membership: |
|

RTF2
Genomic Map