Assay Details
Gene Symbol: LMNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111257.2 | NP_001104727.2 | 8 - 9 | 1722 | 57 | |
XM_005663272.3 | XP_005663329.1 | 1728 | 57 | |||
XM_013996961.2 | XP_013852415.1 | 1822 | 57 | |||
XM_021088439.1 | XP_020944098.1 | 1800 | 57 | |||
XM_021088440.1 | XP_020944099.1 | 2316 | 57 | |||
XM_021088441.1 | XP_020944100.1 | 1754 | 57 | |||
XM_021088442.1 | XP_020944101.1 | 2246 | 57 | |||
XM_021088443.1 | XP_020944102.1 | 1707 | 57 | |||
XM_021088444.1 | XP_020944103.1 | 1734 | 57 | |||
XM_021088445.1 | XP_020944104.1 | 2294 | 57 | |||
XM_021088446.1 | XP_020944105.1 | 2224 | 57 | |||
GenBank mRNA | AK239964.1 | - | 5 - 6 | 770 | 57 | |
AK345149.1 | - | 7 - 8 | 1583 | 57 | ||
AK351165.1 | - | 7 - 8 | 1584 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
