Assay Details
| Gene Symbol: IKBKG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113053.1 | NP_001106524.1 | 1 - 2 | 185 | 65 | |
| XM_005674035.3 | XP_005674092.1 | 385 | 65 | |||
| XM_005674036.3 | XP_005674093.1 | 337 | 65 | |||
| XM_005674040.3 | XP_005674097.1 | 1232 | 65 | |||
| XM_013986571.2 | XP_013842025.1 | 223 | 65 | |||
| XM_021079370.1 | XP_020935029.1 | 351 | 65 | |||
| XM_021079371.1 | XP_020935030.1 | 351 | 65 | |||
| GenBank mRNA | EU258760.1 | - | 1 - 2 | 185 | 65 | |
| EU399821.1 | - | 1 - 2 | 185 | 65 | ||
More Information
Set Membership: |
|

IKBKG
Genomic Map