Assay Details
| Gene Symbol: POU5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113060.1 | NP_001106531.1 | 2 - 3 | 525 | 73 | |
| XM_021097869.1 | XP_020953528.1 | 619 | 73 | |||
| GenBank mRNA | KJ023672.1 | - | 1 - 2 | 552 | 73 | |
More Information
Set Membership: |
|

POU5F1
Genomic Map