Assay Details
| Gene Symbol: SLA-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113704.1 | NP_001107176.1 | 3 - 4 | 738 | 76 | |
| XM_013977620.2 | XP_013833074.2 | 796 | 76 | |||
| GenBank mRNA | AF464006.1 | - | 3 - 4 | 738 | 76 | |
| AF464007.1 | - | 3 - 4 | 738 | 76 | ||
| AF464008.1 | - | 3 - 4 | 738 | 76 | ||
| AF464019.1 | - | 3 - 4 | 738 | 76 | ||
| AF464020.2 | - | 3 - 4 | 700 | 76 | ||
| AK239694.1 | - | 3 - 4 | 719 | 76 | ||
| AK392987.1 | - | 3 - 4 | 723 | 76 | ||
| AY247771.1 | - | 3 - 4 | 738 | 76 | ||
| AY247772.1 | - | 3 - 4 | 738 | 76 | ||
| AY459304.1 | - | 3 - 4 | 738 | 76 | ||
| AY463540.1 | - | 3 - 4 | 698 | 76 | ||
| DQ883215.1 | - | 3 - 4 | 738 | 76 | ||
| DQ883216.1 | - | 3 - 4 | 738 | 76 | ||
| DQ976363.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992502.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992503.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992504.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992505.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992506.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992507.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992508.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992509.1 | - | 3 - 4 | 726 | 76 | ||
| DQ992510.1 | - | 3 - 4 | 726 | 76 | ||
| EU432054.1 | - | 3 - 4 | 719 | 76 | ||
| EU432055.1 | - | 3 - 4 | 719 | 76 | ||
| EU432056.1 | - | 3 - 4 | 719 | 76 | ||
| EU432057.1 | - | 3 - 4 | 719 | 76 | ||
| GU322911.1 | - | 3 - 4 | 704 | 76 | ||
More Information
Set Membership: |
|

SLA-6
Genomic Map