Assay Details
| Gene Symbol: SLA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113706.1 | NP_001107178.1 | 1 - 2 | 147 | 76 | |
| GenBank mRNA | AB215118.1 | - | 1 - 2 | 70 | 76 | |
| AB215119.1 | - | 1 - 2 | 70 | 76 | ||
| AK231578.1 | - | 1 - 2 | 292 | 76 | ||
| AK239665.1 | - | 1 - 2 | 160 | 76 | ||
| AK351169.1 | - | 1 - 2 | 149 | 76 | ||
| AK351896.1 | - | 1 - 2 | 151 | 76 | ||
| AK389739.1 | - | 1 - 2 | 170 | 76 | ||
| AK394576.1 | - | 1 - 2 | 159 | 76 | ||
| AK398000.1 | - | 1 - 2 | 152 | 76 | ||
| AK398178.1 | - | 1 - 2 | 146 | 76 | ||
| AK398242.1 | - | 1 - 2 | 149 | 76 | ||
| AK398994.1 | - | 1 - 2 | 149 | 76 | ||
| AY191779.1 | - | 1 - 2 | 93 | 76 | ||
| AY243102.1 | - | 1 - 2 | 70 | 76 | ||
| AY243106.1 | - | 1 - 2 | 70 | 76 | ||
| AY247781.1 | - | 1 - 2 | 92 | 76 | ||
| AY247782.1 | - | 1 - 2 | 92 | 76 | ||
| AY247783.1 | - | 1 - 2 | 92 | 76 | ||
| AY285926.1 | - | 1 - 2 | 72 | 76 | ||
| AY285928.1 | - | 1 - 2 | 70 | 76 | ||
| AY285929.1 | - | 1 - 2 | 70 | 76 | ||
| AY285933.1 | - | 1 - 2 | 72 | 76 | ||
| AY285935.1 | - | 1 - 2 | 72 | 76 | ||
| AY285936.1 | - | 1 - 2 | 70 | 76 | ||
| AY285937.1 | - | 1 - 2 | 70 | 76 | ||
| AY285938.2 | - | 1 - 2 | 70 | 76 | ||
| AY285939.2 | - | 1 - 2 | 70 | 76 | ||
| AY459302.2 | - | 1 - 2 | 70 | 76 | ||
| DQ298395.1 | - | 1 - 2 | 70 | 76 | ||
| DQ883222.1 | - | 1 - 2 | 92 | 76 | ||
| DQ883223.1 | - | 1 - 2 | 92 | 76 | ||
| DQ883224.1 | - | 1 - 2 | 92 | 76 | ||
| EF143987.1 | - | 1 - 2 | 86 | 76 | ||
| EU432070.1 | - | 1 - 2 | 72 | 76 | ||
| EU432071.1 | - | 1 - 2 | 72 | 76 | ||
| EU722916.1 | - | 1 - 2 | 96 | 76 | ||
| EU722917.1 | - | 1 - 2 | 96 | 76 | ||
| EU722918.1 | - | 1 - 2 | 93 | 76 | ||
| EU728595.1 | - | 1 - 2 | 86 | 76 | ||
| JQ320285.1 | - | 1 - 2 | 70 | 76 | ||
| JQ320286.1 | - | 1 - 2 | 70 | 76 | ||
| JQ361659.1 | - | 1 - 2 | 70 | 76 | ||
| KF026044.1 | - | 1 - 2 | 70 | 76 | ||
| KJ776449.1 | - | 1 - 2 | 70 | 76 | ||
| KJ776450.1 | - | 1 - 2 | 70 | 76 | ||
| KU052691.1 | - | 1 - 2 | 70 | 76 | ||
| KU052692.1 | - | 1 - 2 | 70 | 76 | ||
| KU052693.1 | - | 1 - 2 | 70 | 76 | ||
| KU052694.1 | - | 1 - 2 | 70 | 76 | ||
| KU052695.1 | - | 1 - 2 | 70 | 76 | ||
| KU052696.1 | - | 1 - 2 | 70 | 76 | ||
| KU052697.1 | - | 1 - 2 | 70 | 76 | ||
| KU052698.1 | - | 1 - 2 | 70 | 76 | ||
| KU052699.1 | - | 1 - 2 | 70 | 76 | ||
| KU052700.1 | - | 1 - 2 | 70 | 76 | ||
| KU052701.1 | - | 1 - 2 | 70 | 76 | ||
| KU052702.1 | - | 1 - 2 | 70 | 76 | ||
| KU052703.1 | - | 1 - 2 | 70 | 76 | ||
| KU052704.1 | - | 1 - 2 | 70 | 76 | ||
| M92445.1 | - | 1 - 2 | 179 | 76 | ||
| M93028.1 | - | 1 - 2 | 147 | 76 | ||
More Information
Set Membership: |
|

SLA-DRA
Genomic Map