Assay Details
| Gene Symbol: SLA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113707.1 | NP_001107179.1 | 2 - 3 | 571 | 83 | |
| GenBank mRNA | AK237327.1 | - | 2 - 3 | 564 | 83 | |
| AK239700.1 | - | 2 - 3 | 528 | 83 | ||
| AK389624.1 | - | 1 - 2 | 243 | 83 | ||
| AK394508.1 | - | 2 - 3 | 588 | 83 | ||
| AK398983.1 | - | 2 - 3 | 579 | 83 | ||
| DQ431246.1 | - | 2 - 3 | 352 | 83 | ||
More Information
Set Membership: |
|

SLA-DMB
Genomic Map