Assay Details
| Gene Symbol: SLA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113707.1 | NP_001107179.1 | 4 - 5 | 979 | 73 | |
| GenBank mRNA | AK237327.1 | - | 4 - 5 | 972 | 73 | |
| AK239700.1 | - | 4 - 5 | 936 | 73 | ||
| AK389624.1 | - | 3 - 4 | 651 | 73 | ||
| AK394508.1 | - | 4 - 5 | 996 | 73 | ||
| AK398983.1 | - | 4 - 5 | 987 | 73 | ||
| DQ431246.1 | - | 4 - 5 | 760 | 73 | ||
More Information
Set Membership: |
|

SLA-DMB
Genomic Map