Assay Details
| Gene Symbol: MIC-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114274.1 | NP_001107746.1 | 2 - 3 | 339 | 68 | |
| Gene Symbol: LOC100153163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128306.4 | XP_003128354.1 | 397 | 68 | ||
| XM_013977606.2 | XP_013833060.1 | 412 | 68 | |||
| Gene Symbol: USP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK398518.1 | - | 192 | 68 | ||
More Information
Set Membership: |
|

MIC-2
Genomic Map