Assay Details
| Gene Symbol: CXCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114289.2 | NP_001107761.2 | 3 - 4 | 315 | 152 | |
| GenBank mRNA | AK352281.1 | - | 3 - 4 | 348 | 153 | |
| EU364897.2 | - | 3 - 4 | 315 | 152 | ||
More Information
Set Membership: |
|

CXCL9
Genomic Map