Assay Details
| Gene Symbol: ODC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122983.1 | NP_001116455.1 | 9 - 10 | 1257 | 71 | |
| XM_021085889.1 | XP_020941548.1 | 1699 | 71 | |||
| GenBank mRNA | AB529865.1 | - | 9 - 10 | 1245 | 71 | |
| AK235636.1 | - | 10 - 11 | 1303 | 71 | ||
| AY550057.1 | - | 2 - 3 | 318 | 71 | ||
| EU534186.1 | - | 9 - 10 | 1257 | 71 | ||
More Information
Set Membership: |
|

ODC1
Genomic Map