Assay Details
Gene Symbol: AMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123076.1 | NP_001116548.1 | 10 - 11 | 1681 | 79 | |
XM_005655414.3 | XP_005655471.1 | 1806 | 79 | |||
XM_021088431.1 | XP_020944090.1 | 1912 | 79 | |||
XM_021088432.1 | XP_020944091.1 | 1745 | 79 | |||
GenBank mRNA | AK347120.1 | - | 12 - 13 | 1753 | 79 | |
AK347213.1 | - | 4 - 5 | 556 | 79 | ||
EF634313.2 | - | 10 - 11 | 1681 | 79 | ||
EU545257.1 | - | 10 - 11 | 1677 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
