Assay Details
| Gene Symbol: BLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123084.2 | NP_001116556.1 | 18 - 19 | 3783 | 71 | |
| XM_021097881.1 | XP_020953540.1 | 4042 | 71 | |||
| XM_021097882.1 | XP_020953541.1 | 3897 | 71 | |||
| XM_021097883.1 | XP_020953542.1 | 3528 | 71 | |||
| XM_021097884.1 | XP_020953543.1 | 3920 | 71 | |||
| XM_021097885.1 | XP_020953544.1 | 3726 | 71 | |||
| GenBank mRNA | AB355635.1 | - | 18 - 19 | 3784 | 71 | |
More Information
Set Membership: |
|

BLM
Genomic Map