Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 11 - 12 | 1391 | 122 | |
| GenBank mRNA | EU076594.1 | - | 11 - 12 | 1392 | 122 | |
More Information
Set Membership: |
|

C4A
Genomic Map