Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 32 - 33 | 4279 | 92 | |
| GenBank mRNA | AK232400.1 | - | 3 - 4 | 299 | 92 | |
| AK350147.1 | - | 1 - 2 | 112 | 92 | ||
| AY349423.1 | - | 4 - 5 | 556 | 92 | ||
| EU076594.1 | - | 32 - 33 | 4280 | 92 | ||
More Information
Set Membership: |
|

C4A
Genomic Map