Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 34 - 35 | 4553 | 87 | |
| GenBank mRNA | AK232400.1 | - | 5 - 6 | 573 | 87 | |
| AK350147.1 | - | 3 - 4 | 386 | 87 | ||
| AY349423.1 | - | 6 - 7 | 830 | 87 | ||
| EU076594.1 | - | 34 - 35 | 4554 | 87 | ||
More Information
Set Membership: |
|

C4A
Genomic Map