Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 35 - 36 | 4627 | 63 | |
| GenBank mRNA | AK232400.1 | - | 6 - 7 | 647 | 63 | |
| AK350147.1 | - | 4 - 5 | 460 | 63 | ||
| AY349423.1 | - | 7 - 8 | 904 | 63 | ||
| EU076594.1 | - | 35 - 36 | 4628 | 63 | ||
More Information
Set Membership: |
|

C4A
Genomic Map