Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 37 - 38 | 4815 | 60 | |
| GenBank mRNA | AK232400.1 | - | 8 - 9 | 835 | 60 | |
| AK350147.1 | - | 6 - 7 | 648 | 60 | ||
| AY349423.1 | - | 9 - 10 | 1092 | 60 | ||
| EU076594.1 | - | 37 - 38 | 4816 | 60 | ||
More Information
Set Membership: |
|

C4A
Genomic Map