Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 38 - 39 | 4916 | 98 | |
| GenBank mRNA | AK232400.1 | - | 9 - 10 | 936 | 98 | |
| AK350147.1 | - | 7 - 8 | 749 | 98 | ||
| AY349423.1 | - | 10 - 11 | 1193 | 98 | ||
| EU076594.1 | - | 38 - 39 | 4917 | 98 | ||
More Information
Set Membership: |
|

C4A
Genomic Map