Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 39 - 40 | 5002 | 81 | |
| GenBank mRNA | AK232400.1 | - | 10 - 11 | 1022 | 81 | |
| AK350147.1 | - | 8 - 9 | 835 | 81 | ||
| AY349423.1 | - | 11 - 12 | 1279 | 81 | ||
| EU076594.1 | - | 39 - 40 | 5003 | 81 | ||
More Information
Set Membership: |
|

C4A
Genomic Map