Assay Details
| Gene Symbol: C4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123089.2 | NP_001116561.1 | 40 - 41 | 5137 | 98 | |
| GenBank mRNA | AK232400.1 | - | 11 - 12 | 1157 | 98 | |
| AK350147.1 | - | 9 - 10 | 970 | 98 | ||
| EU076594.1 | - | 40 - 41 | 5138 | 98 | ||
More Information
Set Membership: |
|

C4A
Genomic Map