Assay Details
| Gene Symbol: CTSZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123104.1 | NP_001116576.1 | 2 - 3 | 437 | 139 | |
| GenBank mRNA | AK234297.1 | - | 2 - 3 | 398 | 139 | |
| AK237527.1 | - | 2 - 3 | 363 | 139 | ||
More Information
Set Membership: |
|

CTSZ
Genomic Map