Assay Details
| Gene Symbol: ATAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123123.1 | NP_001116595.1 | 6 - 7 | 540 | 65 | |
| XM_005665740.3 | XP_005665797.1 | 869 | 65 | |||
| XM_005665744.3 | XP_005665801.1 | 869 | 65 | |||
| XM_005665745.3 | XP_005665802.1 | 869 | 65 | |||
| XM_021097944.1 | XP_020953603.1 | 550 | 65 | |||
| GenBank mRNA | AK347360.1 | - | 6 - 7 | 546 | 65 | |
More Information
Set Membership: |
|

ATAT1
Genomic Map