Assay Details
| Gene Symbol: HPCAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123126.1 | NP_001116598.1 | 2 - 3 | 484 | 94 | |
| XM_021085549.1 | XP_020941208.1 | 917 | 94 | |||
| XM_021085550.1 | XP_020941209.1 | 1321 | 94 | |||
| GenBank mRNA | DQ917645.1 | - | 2 - 3 | 484 | 94 | |
More Information
Set Membership: |
|

HPCAL1
Genomic Map