Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123166.1 | NP_001116638.1 | 2 - 3 | 175 | 72 | |
| GenBank mRNA | AK234087.1 | - | 2 - 3 | 265 | 72 | |
More Information
Set Membership: |
|

PPP1R11
Genomic Map