Assay Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123173.1 | NP_001116645.1 | 1 - 2 | 88 | 83 | |
| GenBank mRNA | AK393904.1 | - | 2 - 3 | 145 | 83 | |
| DQ917628.1 | - | 1 - 2 | 88 | 83 | ||
More Information
Set Membership: |
|

RAB1B
Genomic Map