Assay Details
| Gene Symbol: TFAP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123201.1 | NP_001116673.1 | 3 - 4 | 828 | 69 | |
| GenBank mRNA | AK349972.1 | - | 3 - 4 | 816 | 69 | |
More Information
Set Membership: |
|

TFAP2C
Genomic Map