Assay Details
| Gene Symbol: VAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123213.1 | NP_001116685.1 | 3 - 4 | 653 | 97 | |
| XM_021077209.1 | XP_020932868.1 | 7825 | 97 | |||
| GenBank mRNA | AK240055.1 | - | 1 - 2 | 81 | 97 | |
| AK396657.1 | - | 3 - 4 | 570 | 97 | ||
More Information
Set Membership: |
|

VAPB
Genomic Map