Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123221.1 | NP_001116693.1 | 10 - 11 | 1140 | 66 | |
| XM_021097913.1 | XP_020953572.1 | 978 | 66 | |||
| GenBank mRNA | AK233591.1 | - | 10 - 11 | 1075 | 66 | |
| AK234229.1 | - | 10 - 11 | 1166 | 66 | ||
| AK394936.1 | - | 10 - 11 | 1182 | 66 | ||
| AY609916.2 | - | 10 - 11 | 1148 | 66 | ||
| EU442572.1 | - | 10 - 11 | 1140 | 66 | ||
More Information
Set Membership: |
|

HEXA
Genomic Map