Assay Details
| Gene Symbol: ACADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213897.1 | NP_999062.1 | 9 - 10 | 1111 | 61 | |
| GenBank mRNA | D89478.1 | - | 9 - 10 | 1111 | 61 | |
More Information
Set Membership: |
|

ACADL
Genomic Map