Assay Details
| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213902.1 | NP_999067.1 | 3 - 4 | 690 | 71 | |
| XM_021082434.1 | XP_020938093.1 | 1270 | 71 | |||
| GenBank mRNA | Y09400.1 | - | 690 | 71 | ||
More Information
Set Membership: |
|

SLC22A2
Genomic Map