Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213926.1 | NP_999091.1 | 3 - 4 | 319 | 79 | |
| XM_005665624.3 | XP_005665681.1 | 432 | 79 | |||
| GenBank mRNA | AK349129.1 | - | 3 - 4 | 366 | 79 | |
| X14068.1 | - | 319 | 79 | |||
More Information
Set Membership: |
|

PRL
Genomic Map